missing translation for 'onlineSavingsMsg'
Learn More

1,2-Pentanediol 98.0+%, TCI America™

Catalog No. P117825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P117825ML 25 mL
P1178500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P117825ML Supplier TCI America Supplier No. P117825ML

Chemical Identifiers

CAS 5343-92-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00010736
InChI Key WCVRQHFDJLLWFE-UHFFFAOYNA-N
Synonym 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa
PubChem CID 93000
IUPAC Name pentane-1,2-diol
SMILES CCCC(O)CO

Specifications

Color Yellow
Boiling Point 210°C
Quantity 25 mL
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Pentanediol
EINECSNumber (2)-0240
RTECSNumber SA0455000
TSCA Yes