missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methoxyphenyl)piperazine 98.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 25 g |
| UN Number | 3263 |
| Formula Weight | 192.26 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 1-(2-Methoxyphenyl)piperazine |
Sécurité et manipulation
| TSCA | No |