missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)-2-pyrrolidone 98.0+%, TCI America™

Catalog No. H0408500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0408500G 500 g
H040825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0408500G Supplier TCI America Supplier No. H0408500G

Chemical Identifiers

CAS 3445-11-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00014103
InChI Key WDQFELCEOPFLCZ-UHFFFAOYSA-N
PubChem CID 76980
IUPAC Name 1-(2-hydroxyethyl)pyrrolidin-2-one
SMILES C1CC(=O)N(C1)CCO

Specifications

Quantity 500 g
Formula Weight 129.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(2-Hydroxyethyl)-2-pyrrolidone
EINECSNumber (5)-6952
RTECSNumber UY5776500
TSCA Yes