missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedithiol 99.0+%, TCI America™

Catalog No. E0032500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0032500G 500 g
E003225G 25 g
E0032100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0032500G Supplier TCI America Supplier No. E0032500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-63-6
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00004892
InChI Key VYMPLPIFKRHAAC-UHFFFAOYSA-N
Synonym 1,2-ethanedithiol, 1,2-dimercaptoethane, dithioglycol, ethylene mercaptan, ethylenedithiol, 1,2-ethanethiol, ethylenedimercaptan, dithioethyleneglycol, ethylene dithioglycol, s-ethylene dimercaptan
PubChem CID 10902
IUPAC Name ethane-1,2-dithiol
SMILES C(CS)S

Specifications

Color Yellow
Boiling Point 146°C
Quantity 500 g
UN Number 3336
Formula Weight 94.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Ethanedithiol
RTECSNumber KI3325000
TSCA Yes