missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedisulfonic Acid Dihydrate 95.0+%, TCI America™

Catalog No. E04535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04535G 5 g
E045325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04535G Supplier TCI America Supplier No. E04535G

Chemical Identifiers

CAS 110-04-3
Molecular Formula C2H6O6S2
Molecular Weight (g/mol) 190.18
MDL Number MFCD00069901
InChI Key AFAXGSQYZLGZPG-UHFFFAOYSA-N
Synonym 1,2-ethanedisulfonic acid, ethanedisulfonic acid, unii-dl69y31qqv, 1,2-ethanedisulphonic acid, dl69y31qqv, ethane-1,2-disulphonic acid, ethane-1 ,2-disulfonic acid, allergosil, 1,2-ethanedisulfonic acid hydrate, edisylate
PubChem CID 8032
IUPAC Name ethane-1,2-disulfonic acid
SMILES OS(=O)(=O)CCS(O)(=O)=O

Specifications

Melting Point 112°C
Quantity 5 g
UN Number 2585
Formula Weight 190.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Ethanedisulfonic Acid Dihydrate