missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxytetradecane 95.0+%, TCI America™

Catalog No. E0314500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0314500ML 500 mL
E031425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0314500ML Supplier TCI America Supplier No. E0314500ML

Chemical Identifiers

CAS 3234-28-4
Molecular Formula C14H28O
Molecular Weight (g/mol) 212.377
MDL Number MFCD00037524
InChI Key IOHJQSFEAYDZGF-UHFFFAOYSA-N
Synonym 1,2-Tetradecylene Oxide
PubChem CID 18604
IUPAC Name 2-dodecyloxirane
SMILES CCCCCCCCCCCCC1CO1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 500 mL
Formula Weight 212.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxytetradecane
EINECSNumber (2)-0220
TSCA Yes