missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydecane 97.0+%, TCI America™

Catalog No. E0315500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0315500ML 500 mL
E031525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0315500ML Supplier TCI America Supplier No. E0315500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2404-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00005158
InChI Key AAMHBRRZYSORSH-UHFFFAOYNA-N
Synonym 1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
PubChem CID 16993
IUPAC Name 2-octyloxirane
SMILES CCCCCCCCC1CO1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 500 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxydecane
EINECSNumber (2)-0220
TSCA Yes