missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxycyclododecane (mixture of isomers) 95.0+%, TCI America™

Numéro de catalogue. E03225ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E03225ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. E03225ML Fournisseur TCI America Code fournisseur E03225ML
Il en reste null

Spécifications

CAS 286-99-7
Color Yellow
Boiling Point 130°C
Molecular Formula C12H22O
Quantity 5 mL
MDL Number MFCD00005116
Synonym 1,2-epoxycyclododecane, 13-oxabicyclo 10.1.0 tridecane, epoxycyclododecane, cyclododecene oxide, cyclododecene epoxide, cyclododecane epoxide, cyclododecane, 1,2-epoxy, ccris 3754, cyclododecane oxide
InChI Key VLJLXEKIAALSJE-UHFFFAOYNA-N
SMILES C1CCCCCC2OC2CCCC1
IUPAC Name 13-oxabicyclo[10.1.0]tridecane
Molecular Weight (g/mol) 182.31
PubChem CID 9248
Formula Weight 182.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxycyclododecane (mixture of isomers)
Afficher plus de résultats Afficher moins de résultats