missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-5-nitroimidazole 97.0+%, TCI America™

Catalog No. D40815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40815G 5 g
D408125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40815G Supplier TCI America Supplier No. D40815G

Chemical Identifiers

CAS 551-92-8
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00047046
InChI Key IBXPYPUJPLLOIN-UHFFFAOYSA-N
Synonym dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
PubChem CID 3090
IUPAC Name 1,2-dimethyl-5-nitro-1H-imidazole
SMILES CN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 140°C
Color Yellow
Quantity 5 g
Formula Weight 141.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethyl-5-nitroimidazole [for Biochemical Research]
RTECSNumber NI5075000
TSCA No
Recommended Storage Refrigerator