missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-1H-imidazole-5-carbaldehyde, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC25104DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC25104DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

24134-12-1
124.143
XEMFQOKIINBGDI-UHFFFAOYSA-N
3159610
CC1=NC=C(N1C)C=O
C6H8N2O
MFCD06589827
1,2-dimethyl-1h-imidazole-5-carbaldehyde, 2,3-dimethyl-3h-imidazole-4-carbaldehyde, 1h-imidazole-5-carboxaldehyde, 1,2-dimethyl, 1,2-dimethylimidazole-5-carbaldehyde, 1h-imidazole-5-carboxaldehyde,1,2-dimethyl
2,3-dimethylimidazole-4-carbaldehyde

Specifications

1, 2-Dimethyl-1H-imidazole-5-carbaldehyde
96.5
97%
C6H8N2O
MFCD06589827
XEMFQOKIINBGDI-UHFFFAOYSA-N
2,3-dimethylimidazole-4-carbaldehyde
3159610
97%
24134-12-1
100.0
Amber Glass Bottle
1g
1,2-dimethyl-1h-imidazole-5-carbaldehyde, 2,3-dimethyl-3h-imidazole-4-carbaldehyde, 1h-imidazole-5-carboxaldehyde, 1,2-dimethyl, 1,2-dimethylimidazole-5-carbaldehyde, 1h-imidazole-5-carboxaldehyde,1,2-dimethyl
CC1=NC=C(N1C)C=O
124.143
124.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents