missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxy-4-nitrobenzene 99.0+%, TCI America™

Catalog No. D148825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D148825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D148825G Supplier TCI America Supplier No. D148825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 709-09-1
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD00007238
InChI Key YFWBUVZWCBFSQN-UHFFFAOYSA-N
Synonym 3,4-dimethoxynitrobenzene, 4-nitroveratrole, benzene, 1,2-dimethoxy-4-nitro, 4-nitrcveratrole, 4-nitro-1,2-dimethoxybenzene, benzene,2-dimethoxy-4-nitro, 1,2-dimethoxy-4-nitrobenzol, 3,4-dimethoxylnitrobenzene, 3,4-dimethoxy-1-nitrobenzene
PubChem CID 69728
IUPAC Name 1,2-dimethoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 97°C
Color Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 183.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethoxy-4-nitrobenzene