missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxy-4,5-dinitrobenzene, 96%

Catalog No. AAL1170606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1170606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1170606 Supplier Thermo Scientific Chemicals Supplier No. L1170606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3395-03-7 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3395-03-7
MDL Number MFCD00082695

Specifications

Melting Point 131°C to 134°C
Molecular Formula C13H12O4S2
Quantity 5 g
UN Number UN2811
Beilstein 2055812
Synonym bis phenylsulfonyl methane, methylenebis sulphonyl bisbenzene, benzene, 1,1'-methylenebis sulfonyl bis, bis benzenesulfonyl methane, benzenesulfonyl methanesulfonylbenzene, phenylsulfonyl methyl sulfonyl benzene, bisphenylsulfonylmethane, maybridge3_003227, acmc-1cmc8, bis phenylsulphonyl methane
InChI Key QCHNSJNRFSOCLJ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C2=CC=CC=C2
IUPAC Name benzenesulfonylmethylsulfonylbenzene
Molecular Weight (g/mol) 296.36
PubChem CID 76948
Formula Weight 228.16
Percent Purity 96%
Chemical Name or Material 1,2-Dimethoxy-4,5-dinitrobenzene
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only