missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodotetrafluorobenzene, 99%

Numéro de catalogue. AC311690010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC311690010 1 g
AC311690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC311690010 Fournisseur Thermo Scientific Chemicals Code fournisseur 311690010
Il en reste null

CAS: 2708-97-6 | C6F4I2 | 401.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 2708-97-6
Molecular Formula C6F4I2
Molecular Weight (g/mol) 401.87
InChI Key JQBYIZAYQMMVTO-UHFFFAOYSA-N
Synonym 1,2-diiodotetrafluorobenzene, o-diiodotetrafluorobenzene, acmc-1cuq5, 1 2-diiodotetrafluorobenzene
PubChem CID 75923
IUPAC Name 1,2,3,4-tetrafluoro-5,6-diiodobenzene
SMILES C1(=C(C(=C(C(=C1F)I)I)F)F)F

Spécifications

Melting Point 49.0°C to 50.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 401.87
Percent Purity 99%
Chemical Name or Material 1,2-Diiodotetrafluorobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 220-303-3

RUO – Research Use Only