missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodobenzene, 98%

Catalog No. AAB2442503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2442503 1 g
AAB2442506 5 g
AAB2442514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2442503 Supplier Thermo Scientific Chemicals Supplier No. B2442503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-42-9 | C6H4I2 | 329.91 g/mol

1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

Solubility
Slightly miscible with water.

Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatibile with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 615-42-9
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.91
MDL Number MFCD00001038
InChI Key BBOLNFYSRZVALD-UHFFFAOYSA-N
Synonym o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
PubChem CID 11994
IUPAC Name 1,2-diiodobenzene
SMILES IC1=CC=CC=C1I

Specifications

Density 2.524
Boiling Point 282°C
Flash Point >110°C (230°F)
Refractive Index 1.7165
Quantity 1 g
Beilstein 1855315
Sensitivity Light sensitive
Solubility Information Slightly miscible with water.
Formula Weight 329.91
Percent Purity 98%
Chemical Name or Material 1,2-Diiodobenzene
Show More Show Less
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 210-425-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only