missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dihydroxyindan 98.0+%, TCI America™

Numéro de catalogue. D33111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D33111G 1 g
D33115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D33111G Fournisseur TCI America Code fournisseur D33111G
Il en reste null

Identifiants chimiques

CAS 4370-02-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key YKXXBEOXRPZVCC-UHFFFAOYNA-N
Synonym 1,2-Indandiol
PubChem CID 521197
IUPAC Name 2,3-dihydro-1H-indene-1,2-diol
SMILES C1C(C(C2=CC=CC=C21)O)O

Spécifications

Melting Point 158°C
Color White-Yellow
Quantity 1 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dihydroxyindan