missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, 99+%

Catalog No. AAA12775L8
Change view
Click to view available options
Quantity:
500 mL
2500 mL
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12775L8 10 L
AAA12775AP 500 mL
AAA127750F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA12775L8 Supplier Thermo Scientific Chemicals Supplier No. A12775L8
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dichloroethane is used to make vinyl chloride and chlorinated solvents (trichloroethane, trichloroethylene, perchloroethylene, and vinylidene) to remove grease, resins, glue and dirt. It is used as a solvent in the manufacture of polystyrene and SBR latex. It is added to leaded petrol as an anti-knock compound. It is also used in polyvinyl chloride (PVC) pipes, packaging materials, furniture, auto mobile parts, wall coverings and housewares. It is also used generally as an intermediate for other organic chemical compounds and as a solvent. It is a useful solvent for the isomer distribution in the acetylation of benzene and toluene under the influence of aluminum chloride.

Solubility
Miscible with methanol, diethyl ether, n-octanol and acetone. Very slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents, bases and alkali metals.

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35°C
Density 1.256
Quantity 10 L
Flash Point 15°C (59°F)
Refractive Index 1.4448
Linear Formula ClCH2CH2Cl
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Boiling Point 83°C
Solubility Information Miscible with methanol,diethyl ether,n-octanol and acetone. Very slightly miscible with water.
Formula Weight 98.96
Percent Purity ≥99%
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Category H225-H302-H315-H319-H331-H335-H336-H350-H373
Hazard Statement GHS H Statement
H225-H350-H302-H315-H319-H335
Highly flammable liquid and vapour.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P370+P378q-P5
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE
EINECSNumber 203-458-1
RTECSNumber KI0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only