missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020362
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020362 1 mL
111018892 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020362 Supplier Merck Emd Millipore Supplier No. 025621ML
Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 775°F
CAS 107-06-2
Vapor Pressure 87 mmHg (25°C)
Vapor Density 3.4 (20°C, vs. air)
Density 1.256 g/mL (at 25°C)
Flash Point 13°C - Closed Cup
Linear Formula ClCH2 CH2 Cl
MDL Number MFCD00000963
Melting Point -35°C
Boiling Point 83°C
Synonym Ethylene chloride; Ethylene dichloride
Type Analytical Standard
Quantity 1 mL
Formula Weight 98.96
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less