missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene 99.0+%, TCI America™

Catalog No. D1116500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1116500ML 500 mL
D111625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1116500ML Supplier TCI America Supplier No. D1116500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 180°C
Quantity 500 mL
UN Number 1591
Formula Weight 147.00
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichlorobenzene
EINECSNumber (3)-0041
RTECSNumber CZ4500000
TSCA Yes