missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99+%, for spectroscopy

Catalog No. AC222055000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222055000 500 mL Glass bottle
AC222050025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC222055000 Supplier Thermo Scientific Chemicals Supplier No. 222055000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-50-1 | C6H4Cl2 | 147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Alkalinity 0.0005 meq/g max.
Acidity 0.0003 meq/g max.
CAS Min % 99.0
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 at 380nm, 0.05 at 336nm, 0.3 at 300nm, 1 at 296nm
CAS Max % 100.0
Melting Point -15.0°C
Density 1.3000g/mL
Boiling Point 179.0°C to 180.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5520
Linear Formula C6H4Cl2
Quantity 500 mL
Residue after Evaporation 0.0005% max.
Beilstein 05, 201
Specific Gravity 1.3
Merck Index 15, 3065
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Formula Weight 147
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.
May cause
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
EINECSNumber 202-425-9
RTECSNumber CZ4500000
TSCA TSCA

RUO – Research Use Only