missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromoethylene (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. D018625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D018625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D018625G Supplier TCI America Supplier No. D018625G
Only null left

Chemical Identifiers

CAS 540-49-8
Molecular Formula C2H2Br2
Molecular Weight (g/mol) 185.846
MDL Number MFCD00000184
InChI Key UWTUEMKLYAGTNQ-UPHRSURJSA-N
Synonym ethene, 1,2-dibromo-, z, unii-0s4d0712fy, acetylene dibromide, z-1,2-dibromoethene, 1,2-dibromoethylene, cis-dibromoethylene, 1,2-dibromoethylene, 1z, cis-dibromoethene, ethene,2-dibromo, ethylene,2-dibromo
PubChem CID 643776
IUPAC Name (Z)-1,2-dibromoethene
SMILES C(=CBr)Br

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
UN Number 2810
Formula Weight 185.85
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dibromoethylene (cis- and trans- mixture)
RTECSNumber KV7700000
TSCA Yes