missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobenzene, 98%

Catalog No. AAA1476530
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1476530 250 g
AAA1476514 25 g
AAA1476518 50 g
AAA1476522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1476530 Supplier Thermo Scientific Chemicals Supplier No. A1476530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-53-9 | C6H4Br2 | 235.906 g/mol

1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromobenzene is used as a precursor in the preparation of 3-(2-bromophenylthio)-4H-[1]benzopyran-4-one using copper as a reagent. It is involved in the preparation of isoindoloquinazolinones by the carbonylation reaction with 2-aminobenzyl amine using palladium as the catalyst.

Solubility
Miscible with ethanol, ethyl ether, acetone, chloroform, acetic acid, benzene, petroleum ether and tetrachloromethane. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 583-53-9
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.906
MDL Number MFCD00000057
InChI Key WQONPSCCEXUXTQ-UHFFFAOYSA-N
Synonym o-dibromobenzene, dibromobenzene, benzene, 1,2-dibromo, ortho-dibromobenzene, benzene, o-dibromo, benzene, dibromo, unii-52k9w7u6eh, dibromobenzene mixed isomers, 1,2 dibromobenzene, 1,6-dibromobenzene
PubChem CID 11414
ChEBI CHEBI:37152
IUPAC Name 1,2-dibromobenzene
SMILES C1=CC=C(C(=C1)Br)Br

Specifications

Melting Point 4°C to 6°C
Density 1.985
Boiling Point 224°C to 226°C
Flash Point 91°C (195°F)
Odor Strong
Refractive Index 1.611
Quantity 250 g
UN Number UN3082
Beilstein 970241
Solubility Information Miscible with ethanol,ethyl ether,acetone,chloroform,acetic acid,benzene,petroleum ether and tetrachloromethane. Immiscible with water.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobenzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 209-507-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only