missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4,5-dimethylbenzene 98.0+%, TCI America™

Catalog No. D22725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22725G 5 g
D227225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22725G Supplier TCI America Supplier No. D22725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24932-48-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00082743
InChI Key BCIDDURGCAHERU-UHFFFAOYSA-N
PubChem CID 34347
IUPAC Name 1,2-dibromo-4,5-dimethylbenzene
SMILES CC1=CC(Br)=C(Br)C=C1C

Specifications

Melting Point 87°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Synonym 4,5-dibromo-o-xylene, benzene, 1,2-dibromo-4,5-dimethyl, acmc-209gfu, benzene,1,2-dibromo-4,5-dimethyl, 1,2-bis bromanyl-4,5-dimethyl-benzene, 1,2-dibromo-4,5-dimethylbenzene, 1,2-dibromo-4,5-dimethyl-benzene
Boiling Point 278°C
Formula Weight 263.96
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2-Dibromo-4,5-dimethylbenzene