missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylbenzene, 97%

Catalog No. AAL01592MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL01592MD 250 mg
AAL0159203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL01592MD Supplier Thermo Scientific Chemicals Supplier No. L01592MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1159-86-0 | C20H14O2 | 286.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1159-86-0
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00003078
InChI Key OJLABXSUFRIXFL-UHFFFAOYSA-N
Synonym 1,2-dibenzoylbenzene, 1,2-phenylenebis phenylmethanone, 2-benzoylbenzophenone, 2-benzoylphenyl phenyl methanone, methanone, 1,2-phenylenebis phenyl, ortho-dibenzoylbenzene, o-dibenzoylbenzene, methanone,1,1'-1,2-phenylene bis 1-phenyl, phenyl 2-phenylcarbonyl phenyl ketone
PubChem CID 70875
IUPAC Name (2-benzoylphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)C3=CC=CC=C3

Specifications

Melting Point 145°C to 148°C
Quantity 250 mg
Beilstein 789077
Formula Weight 286.33
Percent Purity 97%
Chemical Name or Material 1,2-Dibenzoylbenzene
EINECSNumber 214-597-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only