missing translation for 'onlineSavingsMsg'
Learn More

1,2-Decanediol 99.0+%, TCI America™

Catalog No. D272025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D272025G 25 g
D2720100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D272025G Supplier TCI America Supplier No. D272025G

Chemical Identifiers

CAS 1119-86-4
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00010739
InChI Key YSRSBDQINUMTIF-UHFFFAOYSA-N
Synonym 1,2-decanediol, decylene glycol, decane 1,2-diol, 1,2-dihydroxydecane, acmc-20apfz, acmc-20apjb, pubchem12680, acmc-1bny3
PubChem CID 79141
IUPAC Name decane-1,2-diol
SMILES CCCCCCCCC(CO)O

Specifications

Melting Point 49°C
Color White
Boiling Point 168°C
Quantity 25 g
Formula Weight 174.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Decanediol
EINECSNumber (2)-0240
TSCA No