missing translation for 'onlineSavingsMsg'
Learn More

1,2-Butylene Oxide, MP Biomedicals

Catalog No. ICN201811
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201811 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201811 Supplier MP Biomedicals Inc Supplier No. 0520181180
Only null left

Chemical Identifiers

CAS 106-88-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
InChI Key RBACIKXCRWGCBB-UHFFFAOYSA-N
Synonym 1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
PubChem CID 7834
ChEBI CHEBI:82326
IUPAC Name 2-ethyloxirane
SMILES CCC1CO1

Specifications

Quantity 100 g
Formula Weight 72.11
Chemical Name or Material 1, 2-Butylene Oxide