missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylsulfinyl)ethane 97.0+%, TCI America™

Catalog No. B33525G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33525G Supplier TCI America Supplier No. B33525G

Chemical Identifiers

CAS 6099-21-4
Molecular Formula C14H14O2S2
Molecular Weight (g/mol) 278.38
MDL Number MFCD11846120
InChI Key CSZMCUCIIIAHQD-UHFFFAOYNA-N
PubChem CID 6371710
IUPAC Name [2-(benzenesulfinyl)ethanesulfinyl]benzene
SMILES O=S(CCS(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 278.38
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(phenylsulfinyl)ethane