missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)ethane 97.0+%, TCI America™

Catalog No. B113725G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B113725G 25 g
B113710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B113725G Supplier TCI America Supplier No. B113725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1663-45-2
Molecular Formula C26H24P2
Molecular Weight (g/mol) 398.43
MDL Number MFCD00003047
InChI Key QFMZQPDHXULLKC-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino ethane, diphos, ethylenebis diphenylphosphine, bis diphenylphosphine ethane, bis 1,2-diphenylphosphino ethane, phosphine, 1,2-ethanediylbis diphenyl, phosphine, ethylenebis diphenyl, 1,2-bis diphenylphosphine ethane, 1,2-bis diphenylphosphino-ethane, unii-kl33qe52i4
PubChem CID 74267
ChEBI CHEBI:30669
IUPAC Name [2-(diphenylphosphanyl)ethyl]diphenylphosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 25 g
Formula Weight 398.43
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(diphenylphosphino)ethane