missing translation for 'onlineSavingsMsg'
Learn More

[1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride 96.0+%, TCI America™

Catalog No. B22255G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22255G 5 g
B22251G 1 g
B222525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22255G Supplier TCI America Supplier No. B22255G

Chemical Identifiers

CAS 14647-23-5
Molecular Formula C26H24Cl2NiP2
Molecular Weight (g/mol) 528.02
MDL Number MFCD00013313
InChI Key XXECWTBMGGXMKP-UHFFFAOYSA-L
Synonym 1,2-bis diphenylphosphino ethane nickel ii chloride
PubChem CID 131664338
IUPAC Name [2-(diphenylphosphanyl)ethyl]diphenylphosphane; dichloronickel
SMILES Cl[Ni]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 528.02
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,2-Bis(diphenylphosphino)ethane]nickel(II) Dichloride