missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)benzene 98.0+%, TCI America™

Catalog No. B33725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33725G 5 g
B33721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33725G Supplier TCI America Supplier No. B33725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13991-08-7
Molecular Formula C30H24P2
Molecular Weight (g/mol) 446.47
MDL Number MFCD00014081
InChI Key NFRYVRNCDXULEX-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene
PubChem CID 498379
IUPAC Name [2-(diphenylphosphanyl)phenyl]diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
Formula Weight 446.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene