missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)benzene, 98%

Catalog No. AC363750010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363750010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC363750010 Supplier Thermo Scientific Chemicals Supplier No. 363750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13991-08-7 | C30H24P2 | 446.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13991-08-7
Molecular Formula C30H24P2
Molecular Weight (g/mol) 446.47
MDL Number MFCD00014081
InChI Key NFRYVRNCDXULEX-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene
PubChem CID 498379
IUPAC Name (2-diphenylphosphanylphenyl)-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 184.0°C to 189.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 446.47
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1, 2-Bis(diphenylphosphino)benzene
Show More Show Less

RUO – Research Use Only