missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(dimethylphosphino)ethane 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 180°C |
| Quantity | 1 g |
| UN Number | 1993 |
| Formula Weight | 150.14 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,2-Bis(dimethylphosphino)ethane |
Sécurité et manipulation
| TSCA | No |