missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(chloroacetoxy)ethane 98.0+%, TCI America™
Supplier: TCI America E0107500G
Specifications
| 1,2-Bis(chloroacetoxy)ethane | |
| 41°C | |
| 158°C | |
| MFCD00045277 | |
| Ethylene Glycol Bis(monochloroacetate), Ethylene Glycol Dichloroacetate | |
| C(COC(=O)CCl)OC(=O)CCl | |
| 215.026 | |
| 215.03 | |
| Crystalline Powder |
| 6941-69-1 | |
| White | |
| C6H8Cl2O4 | |
| 500 g | |
| HIIBHBNRMVLLKH-UHFFFAOYSA-N | |
| 2-(2-chloroacetyl)oxyethyl 2-chloroacetate | |
| 81365 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6941-69-1 | |
| 215.026 | |
| HIIBHBNRMVLLKH-UHFFFAOYSA-N | |
| 81365 | |
| C(COC(=O)CCl)OC(=O)CCl |
| C6H8Cl2O4 | |
| MFCD00045277 | |
| Ethylene Glycol Bis(monochloroacetate), Ethylene Glycol Dichloroacetate | |
| 2-(2-chloroacetyl)oxyethyl 2-chloroacetate |
Safety and Handling
TSCA : No