missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(chloroacetoxy)ethane 98.0+%, TCI America™

Catalog No. p-7128842
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E010725G 25 g
E0107500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E010725G Supplier TCI America Supplier No. E010725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6941-69-1
Molecular Formula C6H8Cl2O4
Molecular Weight (g/mol) 215.026
MDL Number MFCD00045277
InChI Key HIIBHBNRMVLLKH-UHFFFAOYSA-N
Synonym Ethylene Glycol Bis(monochloroacetate), Ethylene Glycol Dichloroacetate
PubChem CID 81365
IUPAC Name 2-(2-chloroacetyl)oxyethyl 2-chloroacetate
SMILES C(COC(=O)CCl)OC(=O)CCl

Specifications

Melting Point 41°C
Color White
Boiling Point 158°C
Quantity 25 g
Formula Weight 215.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(chloroacetoxy)ethane