accessibility menu, dialog, popup

UserName

ClOOOOCl

CAS RN 6941-69-1

IUPAC Name: 2-[(2-chloroacetyl)oxy]ethyl 2-chloroacetate
Synonyms: 2-[(2-chloroacetyl)oxy]ethyl 2-chloroacetate
Molecular Weight (g/mol): 215.03
Molecular Formula: C6H8Cl2O4
InChi Key: HIIBHBNRMVLLKH-UHFFFAOYSA-N
SMILES: ClCC(=O)OCCOC(=O)CCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results