missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene 98.0+%, TCI America™

Catalog No. p-7124132
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56051G 1 g
B56055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56051G Supplier TCI America Supplier No. B56051G
Only null left

Chemical Identifiers

CAS 222166-46-3
Molecular Formula C32H20Br4N2
Molecular Weight (g/mol) 752.142
InChI Key ZHQHEWRIQYHYBQ-UHFFFAOYSA-N
Synonym 9,9′-[1,2-Phenylenebis(methylene)]bis(3,6-dibromo-9H-carbazole)
PubChem CID 132274867
IUPAC Name 3,6-dibromo-9-[[2-[(3,6-dibromocarbazol-9-yl)methyl]phenyl]methyl]carbazole
SMILES C1=CC=C(C(=C1)CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)CN5C6=C(C=C(C=C6)Br)C7=C5C=CC(=C7)Br

Specifications

Melting Point 319°C
Quantity 1 g
Formula Weight 752.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene