missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-aminoethoxy)ethane 98.0+%, TCI America™

Catalog No. B143125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B143125G 25 g
B1431100G 100 g
B1431500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B143125G Supplier TCI America Supplier No. B143125G

Chemical Identifiers

CAS 929-59-9
Molecular Formula C6H16N2O2
Molecular Weight (g/mol) 148.206
MDL Number MFCD00040474
InChI Key IWBOPFCKHIJFMS-UHFFFAOYSA-N
Synonym 1,8-diamino-3,6-dioxaoctane, 1,2-bis 2-aminoethoxy ethane, 2,2'-ethylenedioxy bis ethylamine, unii-r41zt4uf34, 2,2'-ethane-1,2-diylbis oxy diethanamine, 3,6-dioxaoctamethylenediamine, 2-2-2-aminoethoxy ethoxy ethanamine, 3,6-dioxaoctane-1,8-diamine, ethylene glycol bis 2-aminoethyl ether
PubChem CID 70248
IUPAC Name 2-[2-(2-aminoethoxy)ethoxy]ethanamine
SMILES C(COCCOCCN)N

Specifications

Color Yellow
Boiling Point 123°C
Quantity 25 g
UN Number 2735
Formula Weight 148.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(2-aminoethoxy)ethane
EINECSNumber (2)-0312
TSCA Yes