missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene 98.0+%, TCI America™

Catalog No. B2629100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2629100MG 100 mg
B26291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2629100MG Supplier TCI America Supplier No. B2629100MG
Only null left

Chemical Identifiers

CAS 172612-67-8
Molecular Formula C29H22F6S2
Molecular Weight (g/mol) 548.605
InChI Key DYZAFEDVNIEMEL-UHFFFAOYSA-N
PubChem CID 10721339
IUPAC Name 3-[2-(2,4-dimethyl-5-phenylthiophen-3-yl)-3,3,4,4,5,5-hexafluorocyclopenten-1-yl]-2,4-dimethyl-5-phenylthiophene
SMILES CC1=C(SC(=C1C2=C(C(C(C2(F)F)(F)F)(F)F)C3=C(SC(=C3C)C4=CC=CC=C4)C)C)C5=CC=CC=C5

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 100 mg
Formula Weight 548.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene