missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene (purified by sublimation) 99.0+%, TCI America™

Catalog No. B5604100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5604100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5604100MG Supplier TCI America Supplier No. B5604100MG
Only null left

Chemical Identifiers

CAS 172612-67-8
Molecular Formula C29H22F6S2
Molecular Weight (g/mol) 548.605
InChI Key DYZAFEDVNIEMEL-UHFFFAOYSA-N
PubChem CID 10721339
IUPAC Name 3-[2-(2,4-dimethyl-5-phenylthiophen-3-yl)-3,3,4,4,5,5-hexafluorocyclopenten-1-yl]-2,4-dimethyl-5-phenylthiophene
SMILES CC1=C(SC(=C1C2=C(C(C(C2(F)F)(F)F)(F)F)C3=C(SC(=C3C)C4=CC=CC=C4)C)C)C5=CC=CC=C5

Specifications

Melting Point 132°C
Color Blue-White
Quantity 100 mg
Formula Weight 548.61
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene (purified by sublimat