missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedimethanol, 97%

Catalog No. AAB2300803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2300803 1 g
AAB2300806 5 g
AAB2300814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2300803 Supplier Thermo Scientific Chemicals Supplier No. B2300803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

1,2-Benzenedimethanol is a raw material used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.

Solubility
Soluble in water, ether, ethanol, benzene.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

Melting Point 59°C to 64°C
Quantity 1 g
Beilstein 508775
Solubility Information Soluble in water,ether,ethanol,benzene.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedimethanol
EINECSNumber 210-293-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only