missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. A088425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A088425G Supplier TCI America Supplier No. A088425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7154-73-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00003182
InChI Key WRXNJTBODVGDRY-UHFFFAOYSA-N
Synonym 1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
PubChem CID 1344
IUPAC Name 2-pyrrolidin-1-ylethanamine
SMILES C1CCN(C1)CCN

Specifications

Color Yellow
Boiling Point 161°C
Quantity 25 g
UN Number 2733
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Aminoethyl)pyrrolidine