missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Aminoethyl)-4-methylpiperazine, 97+%

Catalog No. AAH52371MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52371MD 250 mg
AAH5237103 1 g
AAH5237106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52371MD Supplier Thermo Scientific Chemicals Supplier No. H52371MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 934-98-5 | C7H17N3 | 143.234 g/mol

1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 934-98-5
Molecular Formula C7H17N3
Molecular Weight (g/mol) 143.234
MDL Number MFCD03701701
InChI Key GOWUDHPKGOIDIX-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl ethanamine, 4-methyl-1-piperazineethanamine, 2-4-methylpiperazin-1-yl ethylamine, 1-piperazineethanamine, 4-methyl, 2-4-methyl-piperazin-1-yl-ethylamine, 4-methylpiperazine-1-ethylamine, 2-4-methylpiperazin-1-yl ethan-1-amine, 1-2-aminoethyl-4-methyl-piperazine, 1-2-aminoethyl-4-methylpiperazine, 1-n-aminoethyl-4-methyl piperazine
PubChem CID 70284
ChEBI CHEBI:84290
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanamine
SMILES CN1CCN(CC1)CCN

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 143.43
Percent Purity ≥97%
Chemical Name or Material 1-(2-Aminoethyl)-4-methylpiperazine
Hazard Category H302+H312-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 213-296-3
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only