missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Dimethylphenoxy)-2-propanone 95.0+%, TCI America™

Catalog No. D51215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51215G 5 g
D51211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51215G Supplier TCI America Supplier No. D51215G
Only null left

Chemical Identifiers

CAS 53012-41-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00130266
InChI Key XDJULAUHYAJQBU-UHFFFAOYSA-N
Synonym (2,6-Dimethylphenoxy)acetone
PubChem CID 104378
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-one
SMILES CC1=C(C(=CC=C1)C)OCC(=O)C

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 113°C
Formula Weight 178.23
Percent Purity ≥95.0% (GC)
Chemical Name or Material 1-(2,6-Dimethylphenoxy)-2-propanone