missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-D-mannitol 97.0+%, TCI America™

Catalog No. D20245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20245G 5 g
D202425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20245G Supplier TCI America Supplier No. D20245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1707-77-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00003211
InChI Key ODYBCPSCYHAGHA-ZYUZMQFOSA-N
Synonym 1,2:5,6-di-o-isopropylidene-d-mannitol, d-mannitol diacetonide, 1,2:5,6-bis-o-1-methylethylidene-d-mannitol, d-mannitol 1,2:5,6-bis-acetonide, 1s,2s-1,2-bis 4r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1s,2s-1,2-bis r-2,2-dimethyl-1,3-dioxolan-4-yl ethane-1,2-diol, 1,2,5,6-di-o-isopropylidene-d-mannitol, mannitol diacetonide, diacetone-d-mannitol, zlchem 213
PubChem CID 96011
IUPAC Name (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol
SMILES CC1(OCC(O1)C(C(C2COC(O2)(C)C)O)O)C

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Formula Weight 262.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2:5,6-Di-O-isopropylidene-D-mannitol