missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,2,4-Trimethylbenzene

Numéro de catalogue. T736012L
Encompass
Modifier l'affichage
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T736012L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. T736012L Fournisseur Sigma Aldrich Code fournisseur T736012L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2,4-Trimethylbenzene

Spécifications

CAS 95-63-6
Melting Point -44°C (lit.)
Density 0.876 g/mL (at 20°C (literature))
Boiling Point 168°C (lit.)
Molecular Formula C9H12
Refractive Index n20/D 1.504 (literature)
Linear Formula C6H3(CH3)3
Quantity 2 L
MDL Number MFCD00008527
Synonym Pseudocumene
Molecular Weight (g/mol) 120.19
Percent Purity 98%
Afficher plus de résultats Afficher moins de résultats