missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, OmniSolv™ MilliporeSigma™

Catalog No. MTX1056P1
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX1056P1 4 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX1056P1 Supplier Technidata Supplier No. TX1056P1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Spectrophotometry

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Melting Point 17°C
Color Colorless
Quantity 4 L
Odor Aromatic
Packaging Poly Coated Glass Bottle
Physical Form Liquid
Vapor Pressure 0.026 kPa (0.195016576mm Hg) [20°C]
UN Number UN 2321
Boiling Point 213.5°C
Solubility Information Water: Partially soluble
Formula Weight 181.44
Percent Purity ≥99.0%
Grade ACS
Chemical Name or Material 1,2,4-Trichlorobenzene
Show More Show Less
DOTInformation DOT Class: 6.1, Packaging Group: III
Recommended Storage Room Temperature