missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, 99.0% min., ACS, MilliporeSigma™

Catalog No. MTX106030
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX106030 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX106030 Supplier Technidata Supplier No. TX106030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-82-1
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
InChI Key PBKONEOXTCPAFI-UHFFFAOYSA-N
Synonym benzene, 1,2,4-trichloro, unsym-trichlorobenzene, hostetex l-pec, trojchlorobenzen, 1,2,4-trichlorbenzol, 1,2,4-trichlorobenzol, 1,3,4-trichlorobenzene, trichlorobenzene a, 1,2,5-trichlorobenzene, as-trichlorobenzene
PubChem CID 13
ChEBI CHEBI:28222
IUPAC Name 1,2,4-trichlorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)Cl

Specifications

Color Colorless
Density 1.45g/cm3 (20°C)
Quantity 200 L
Explosion Limit 2.5 to 6.6%(V)
Ignition Point 571°C
Physical Form Liquid
Assay Percent Range 99.0% min.
Vapor Pressure 1.3 hPa (20°C)
Formula Weight 181.45
Chemical Name or Material 1, 2, 4-Trichlorobenzene, For Synthesis
Recommended Storage Store at +15°C to +25°C