missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazole-1-carboximidamide Hydrochloride 98.0+%, TCI America™

Catalog No. T31245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31245G 5 g
T31241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31245G Supplier TCI America Supplier No. T31245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19503-26-5
Molecular Formula C3H6ClN5
Molecular Weight (g/mol) 147.566
MDL Number MFCD03095468
InChI Key JDDXNENZFOOLTP-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboxamidine monohydrochloride, 1h-1,2,4-triazole-1-carboxamidine hydrochloride, 1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboximidamide, monohydrochloride, c3h5n5.clh, ksc496s5d, 1,2,4-triazolecarboxamidine, chloride, 1-amidino-1,2,4-triazole hydrochloride, 1-amidino-1h-1,2,4-triazole hydrochloride
PubChem CID 16218515
IUPAC Name 1,2,4-triazole-1-carboximidamide;hydrochloride
SMILES C1=NN(C=N1)C(=N)N.Cl

Specifications

Melting Point 199°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 147.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazole-1-carboximidamide Hydrochloride