missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrabromobenzene 97.0+%, TCI America™

Catalog No. T00315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00315G 5 g
T003125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T00315G Supplier TCI America Supplier No. T00315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 636-28-2
Molecular Formula C6H2Br4
Molecular Weight (g/mol) 393.70
MDL Number MFCD00000063
InChI Key QCKHVNQHBOGZER-UHFFFAOYSA-N
PubChem CID 12486
IUPAC Name 1,2,4,5-tetrabromobenzene
SMILES BrC1=CC(Br)=C(Br)C=C1Br

Specifications

Melting Point 180°C
Quantity 5 g
Synonym benzene, 1,2,4,5-tetrabromo, 2,3,5,6-tetrabromobenzene, unii-m25dk66lcz, m25dk66lcz, pubchem4448, 1,4,5-tetrabromobenzene, 2,5,6-tetrabromobenzene, acmc-1b6z1, benzene,2,4,5-tetrabromo, benzene,1,2,4,5-tetrabromo
Formula Weight 393.70
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Tetrabromobenzene