missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Benzenetetracarboxylic acid, 96%

Catalog No. AC105315000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105315000 500 g Plastic bottle
AC105311000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105315000 Supplier Thermo Scientific Chemicals Supplier No. 105315000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-05-4 | C10H6O8 | 254.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-05-4
Molecular Formula C10H6O8
Molecular Weight (g/mol) 254.15
MDL Number MFCD00002471
InChI Key CYIDZMCFTVVTJO-UHFFFAOYSA-N
Synonym pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
PubChem CID 6961
ChEBI CHEBI:45165
IUPAC Name benzene-1,2,4,5-tetracarboxylic acid
SMILES C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 281.0°C to 284.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula C6H2(CO2H)4
Quantity 500 g
Beilstein 09, 997
Merck Index 15, 8116
Solubility Information Solubility in water: 1.5g/100mL water (20°C). Other solubilities: 30g/100mL water (100°C),freely soluble in alcohol
Formula Weight 254.15
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4,5-Benzenetetracarboxylic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-879-5
RTECSNumber DB9275000
TSCA TSCA

RUO – Research Use Only