missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trimethylbenzene 80.0+%, TCI America™

Catalog No. T04685ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04685ML 5 mL
T046825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04685ML Supplier TCI America Supplier No. T04685ML

Chemical Identifiers

CAS 526-73-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008520
InChI Key FYGHSUNMUKGBRK-UHFFFAOYSA-N
Synonym hemimellitene, hemellitol, hemimellitol, trimethylbenzene, benzene, 1,2,3-trimethyl, benzene, trimethyl, unii-zk4r7uph6r, 1,2,3-trimethylbenzene, zk4r7uph6r, 1,3-trimethylbenzene
PubChem CID 10686
ChEBI CHEBI:34037
IUPAC Name 1,2,3-trimethylbenzene
SMILES CC1=C(C(=CC=C1)C)C

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Trimethylbenzene
EINECSNumber (3)-0007&(3)-3427
RTECSNumber DC3300000
TSCA Yes